2-Hydroxy-3-methoxy-5-nitrobenzaldehyd ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

A14360.03 A14360.06 A14360.14
A14360.03EA 30.7 EUR
A14360.03 A14360.06 A14360.14
2-Hydroxy-3-methoxy-5-nitrobenzaldehyd ≥98%
2-Hydroxy-3-methoxy-5-nitrobenzaldehyde

Purity: 98% CAS No.: 17028-61-4 Molecular Formula: C8H7NO5 Formula Weight: 197.15 Melting Point: 138-142° Beilstein Reference: 613271

Formel: C₈H₇NO₅
Molecular Weight: 197.15 g/mol
Schmelzpunkt: 138…142 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00017033
CAS-Nummer: 17028-61-4

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR