Aminoacetaldehyddimethylacetal 99%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
2,2-Dimethoxyethanamine
A15498.14
A15498.22
A15498.36
A15498.14EA
42
EUR
A15498.14
A15498.22
A15498.36
Aminoacetaldehyddimethylacetal 99%
Aminoacetaldehyddimethylacetal
Purity: 99%
CAS No.: 22483-09-6
Molecular Formula: H2NCH2CH(OCH3)2
Formula Weight: 105.14
Flash Point: 41°(106°F)
Refractive Index: 1.4170
Boiling Point: 135-138°
Beilstein Reference: 741868
UN No.: 2733
MDL No.: MFCD00008135
Formel:
C₄H₁₁NO₂ Molecular Weight: 105,14 g/mol Siedepunkt: 135…138 °C Dichte: 0,973 Flammpunkt: 41 °C (106 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008135 CAS-Nummer: 22483-09-6 EINECS: 245-026-5 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...