1,3-Dibrom-5,5-dimethylhydantoin ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 1,3-Dibromo-5,5-dimethylimidazolidine-2,4-dione

A15510.0E A15510.22 A15510.36 AB115290.0100
A15510.0EEA 271 EUR
A15510.0E A15510.22 A15510.36 ABCRAB115290.0100
1,3-Dibrom-5,5-dimethylhydantoin ≥98%
1,3-Dibrom-5,5-dimethylhydantoin

Purity: 98% CAS No.: 77-48-5 Molecular Formula: C5H6Br2N2O2 Formula Weight: 285.93 Flash Point: 155°(311°F) Melting Point: ca 198° dec. Beilstein Reference: 146024 UN No.: 3085 MDL No.: MFCD00003189

Formel: C₅H₆Br₂N₂O₂
Molecular Weight: 285.92 g/mol
Schmelzpunkt: ∼ 198 °C (Zers.)
Flammpunkt: 155 °C (311 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003189
CAS-Nummer: 77-48-5
EINECS: 201-030-9
ADR: 5.1 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR