N-Methylpropylamin 97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
N-Methyl-1-propylamine
A16903.03
A16903.06
A16903.14
A16903.03EA
27.4
EUR
A16903.03
A16903.06
A16903.14
N-Methylpropylamin 97%
N-Methylpropylamin
Purity: 97%
CAS No.: 627-35-0
Molecular Formula: CH3NHCH2CH2CH3
Formula Weight: 73.14
Flash Point: -1.1°(30°F)
Odour: Amine-like
Boiling Point: 62-64°
UN No.: 2733
MDL No.: MFCD00009361
Formel:
C₄H₁₁N Molecular Weight: 73,14 g/mol Siedepunkt: 62…64 °C Dichte: 0,713 Flammpunkt: –1,1 °C (30 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009361 CAS-Nummer: 627-35-0 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...