5,5-Diphenylhydantoin ≥99%

Lieferant: Thermo Scientific
Gefahr

A17013.14 A17013.22 A17013.36
A17013.14EA 23.2 EUR
A17013.14 A17013.22 A17013.36
5,5-Diphenylhydantoin ≥99%
5,5-Diphenylhydantoin

Purity: 99% CAS No.: 57-41-0 Molecular Formula: C15H12N2O2 Formula Weight: 252.27 Melting Point: 293-298° Beilstein Reference: 384532 Merck Reference: 14,7322 MDL No.: MFCD00005264

Formel: C₁₅H₁₂N₂O₂
Molecular Weight: 252,27 g/mol
Schmelzpunkt: 293…298 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005264
CAS-Nummer: 57-41-0
EINECS: 200-328-6
Merck Index: 14,7322

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR