Isonicotinaldehydoxim ≥97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
4-Pyridinecarbaldehyde oxime, Pyridine-4-carboxaldoxime
A17371.14
A17371.22
A17371.14EA
54.5
EUR
A17371.14
A17371.22
Isonicotinaldehydoxim ≥97%
Isonicotinaldehydoxim
Purity: 97%
CAS No.: 696-54-8
Molecular Formula: C6H6N2O
Formula Weight: 122.13
Melting Point: 130-133°
Beilstein Reference: 2079
Formel:
C₆H₆N₂O Molecular Weight: 122,13 g/mol Schmelzpunkt: 130…133 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006446 CAS-Nummer: 696-54-8 EINECS: 211-796-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...