(R)-(-)-β-Aminophenethylalkohol ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: D-Phenylglycinol, (R)-(-)-2-Phenylglycinol

A19030.03 A19030.06 A19030.14
A19030.03EA 30 EUR
A19030.03 A19030.06 A19030.14
(R)-(-)-β-Aminophenethylalkohol ≥98%
(R)-(-)-β-Aminophenethylalkohol

Purity: 98% CAS No.: 56613-80-0 Molecular Formula: C8H11NO Formula Weight: 137.18 Melting Point: 76-78° Beilstein Reference: 2935848

Formel: C₈H₁₁NO
Molecular Weight: 137,18 g/mol
Schmelzpunkt: 76…78 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00008062
CAS-Nummer: 56613-80-0
EINECS: 260-287-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR