L(+)-α-Phenylglycin ≥98%
Lieferant: Thermo Scientific
Synonyme:
H-Phg-OH, L-(+)-2-Phenylglycine
A19360.14
A19360.22
A19360.36
A19360.14EA
44.3
EUR
A19360.14
A19360.22
A19360.36
L(+)-α-Phenylglycin ≥98%
L(+)-α-Phenylglycin
Purity: 98+%
CAS No.: 2935-35-5
Molecular Formula: C8H9NO2
Formula Weight: 151.17
Melting Point: 288° dec.
Beilstein Reference: 2208675
Merck Reference: 14,7291
Optical Rotation: +156° (c=1 in 1N HCl)
MDL No.: MFCD00064403
Formel:
C₈H₉NO₂ Molecular Weight: 151,16 g/mol Schmelzpunkt: 288 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064403 CAS-Nummer: 2935-35-5 EINECS: 220-909-8 Merck Index: 14,7291 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...