Ethyl-2-chlornicotinat ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Ethyl 2-chloropyridine-3-carboxylate, Ethyl 2-Chloro-3-pyridinecarboxylate, Ethyl 2-chloronicotinate
B20359.09
B20359.18
B20359.09EA
55.6
EUR
B20359.09
B20359.18
Ethyl-2-chlornicotinat ≥98%
Ethyl-2-chlornicotinat
Purity: 98%
CAS No.: 1452-94-4
Molecular Formula: C8H8ClNO2
Formula Weight: 185.61
MDL No.: MFCD00674129
Formel:
C₈H₈ClNO₂ Molecular Weight: 185,61 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00674129 CAS-Nummer: 1452-94-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...