Methyl-5-bromnicotinat ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

B20908.06 B20908.14
B20908.06EA 54.1 EUR
B20908.06 B20908.14
Methyl-5-bromnicotinat ≥97%
Methyl-5-bromnicotinat

Purity: 97% CAS No.: 29681-44-5 Molecular Formula: C7H6BrNO2 Formula Weight: 216.04 Melting Point: 96-98° MDL No.: MFCD00173668

Formel: C₇H₆BrNO₂
Schmelzpunkt: 96…98 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00173668
CAS-Nummer: 29681-44-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR