Cystaminiumdichlorid ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Cystaminium dichloride, 2,2′-Diaminodiethyl disulphide dihydrochloride, 2,2'-Disulphanediyldiethanamine dihydrochloride, Bis(2-aminoethyl) disulphide dihydrochloride, Decarboxycystine dihydrochloride, Cystamine (dihydrochloride), {2-[(2-Aminoethyl)disulphanyl]ethyl}amine dihydrochloride, 2,2′-Dithiobis(ethylamine) dihydrochloride

B22873.14 B22873.22
B22873.14EA 38.1 EUR
B22873.14 B22873.22
Cystaminiumdichlorid ≥97%
Cystaminiumdichlorid

Cystamine dihydrochloride acts as an anti-infective agent, which is used in the treatment of urinary tract infections. It is also used as a radiation-protective agent that interferes with sulfhydryl enzymes. Further, it serves as a heparin antagonist and sulfhydryl modifying reagent. In addition to this, it is used as an inhibitor of TGase.


Purity: 97+% CAS No.: 56-17-7 Molecular Formula: C4H12N2S2.2HCl Formula Weight: 225.20 Odour: Odourless Melting Point: 217-220° Beilstein Reference: 3616850 Merck Reference: 14,2776 MDL No.: MFCD00012905

Formel: C₄H₁₂N₂S₂·2HCl
Molecular Weight: 225.20 g/mol
Schmelzpunkt: 217…220 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012905
CAS-Nummer: 56-17-7
EINECS: 200-260-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR