1-(2-Chlorethyl)piperidiniumchlorid ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: N-(2-Chloroethyl)-piperidine hydrochloride, 1-(2-Chloroethyl)piperidine hydrochloride

B22985.18 B22985.30
B22985.18EA 28.5 EUR
B22985.18 B22985.30
1-(2-Chlorethyl)piperidiniumchlorid ≥98%
1-(2-Chlorethyl)piperidiniumchlorid

Purity: 98% CAS No.: 2008-75-5 Molecular Formula: C7H14ClN.HCl Formula Weight: 184.11 Melting Point: 229-235° Beilstein Reference: 3910268 UN No.: 2928 MDL No.: MFCD00012837

Formel: C₇H₁₅Cl₂N
Molecular Weight: 184.11 g/mol
Schmelzpunkt: 229…235 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012837
CAS-Nummer: 2008-75-5
EINECS: 217-920-5
UN: 2928
ADR: 6.1 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR