Pyridin-2-aldoxim methochlorid ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

B23890.06 B23890.14
B23890.06EA 31.1 EUR
B23890.06 B23890.14
Pyridin-2-aldoxim methochlorid ≥97%
Pyridin-2-aldoxim methochlorid

Purity: 97% CAS No.: 51-15-0 Molecular Formula: C7H9ClN2O Formula Weight: 172.62 Melting Point: ca 223° dec. Beilstein Reference: 4163981 Merck Reference: 14,7703 MDL No.: MFCD00011981

Formel: C₇H₉ClN₂O
Schmelzpunkt: ∼ 223 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011981
CAS-Nummer: 51-15-0
EINECS: 200-080-9
Merck Index: 14,7703

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR