Aminoguanidiniumhydrogencarbonat ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Aminoguanidine hydrogen carbonate, Aminoguanidine bicarbonate, 1-Aminoguanidinium hydrogen carbonate, Aminoguanidine hydrogencarbonate, Guanylhydrazine hydrogencarbonate
B24696.0E
B24696.22
B24696.36
B24696.0EEA
187
EUR
B24696.0E
B24696.22
B24696.36
Aminoguanidiniumhydrogencarbonat ≥98%
Aminoguanidinium hydrogen carbonate
Purity: 98+%
CAS No.: 2582-30-1
Molecular Formula: CH6N4.H2CO3
Formula Weight: 136.11
Melting Point: ca 170° dec.
Beilstein Reference: 3569869
UN No.: 3077
MDL No.: MFCD00012949
Formel:
C₂H₈N₄O₃ Molecular Weight: 136,11 g/mol Schmelzpunkt: ∼ 170 °C (Zersetzung) Dichte: 1,600 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012949 CAS-Nummer: 2582-30-1 EINECS: 219-956-7 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...