Thallium(I)bromid ≥99,999% (Metall-Basis)

Lieferant: Thermo Scientific
Gefahr

Synonyme: Thalliumbromid

11841.14 11841.22
11841.14EA 160 EUR
11841.14 11841.22
Thallium(I)bromid ≥99,999% (Metall-Basis)
Thallium(I)bromid

Purity: 99.999% CAS No.: 7789-40-4 Molecular Formula: TlBr Formula Weight: 284.28 Physical Form: Powder Density: 7.5 Melting Point: ca 460° Solubility: Practically insoluble in water Merck Reference: 14,9258 UN No.: 1707 MDL No.: MFCD00011273

Formel: BrTl
Molecular Weight: 284,29 g/mol
Siedepunkt: 815 °C (1013 hPa)
Schmelzpunkt: ∼460 °C
Dichte: 7,5
Lagertemperatur: Raumtemperatur
MDL: MFCD00011273
CAS-Nummer: 7789-40-4
EINECS: 232-163-0
UN: 1707
ADR: 6.1,II
Merck Index: 14,9258

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR