(±)-4-Chlorstyroloxid ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-(4-Chlorophenyl)oxirane
H26516.06
H26516.14
H26516.06EA
87.3
EUR
H26516.06
H26516.14
(±)-4-Chlorstyroloxid ≥98%
(±)-4-Chlorstyroloxid
Purity: 98%
CAS No.: 2788-86-5
Molecular Formula: C8H7ClO
Formula Weight: 154.59
Flash Point:
Formel:
C₈H₇ClO Molecular Weight: 154,6 g/mol Siedepunkt: 230 °C Dichte: 1,283 Flammpunkt: >100 °C (212 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00956179 CAS-Nummer: 2788-86-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...