N-Methylallylamin 96%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
N-Allylmethylamine
H27551.06
H27551.03
H27551.06EA
119
EUR
H27551.06
H27551.03
N-Methylallylamin 96%
N-Methylallylamin
Purity: 96%
CAS No.: 627-37-2
Molecular Formula: CH3NHCH2CH=CH2
Formula Weight: 71.12
Storage Temperature: 2°C to 8°C
Flash Point: -8°(-18°F)
Refractive Index: 1.412
Boiling Point: 61-62°
UN No.: 2733
MDL No.: MFCD00008641
Formel:
C₄H₉N Molecular Weight: 71,12 g/mol Siedepunkt: 61…62 °C Dichte: 0,748 Flammpunkt: –8 °C (–18 °F) |
MDL:
MFCD00008641 CAS-Nummer: 627-37-2 EINECS: 210-996-0 UN: 3286 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...