Methyl-6-aminonicotinat 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Methyl 6-aminonicotinate, 6-Aminonicotinic acid methyl ester
H27575.06
H27575.03
H27575.06EA
84
EUR
H27575.06
H27575.03
Methyl-6-aminonicotinat 97%
Methyl-6-aminonicotinat
Purity: 97%
CAS No.: 36052-24-1
Molecular Formula: C7H8N2O2
Formula Weight: 152.15
Melting Point: 158-161°
MDL No.: MFCD00797844
Formel:
C₇H₈N₂O₂ Molecular Weight: 152,15 g/mol Schmelzpunkt: 158…161 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00797844 CAS-Nummer: 36052-24-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...