3-Aminoisonicotinsäure 95%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3-Aminoisonicotinic Acid, 3-Aminopyridine-4-carboxylic acid
H28714.06
H28714.03
H28714.06EA
665
EUR
H28714.06
H28714.03
3-Aminoisonicotinsäure 95%
3-Aminoisonicotinsäure
Purity: 95%
CAS No.: 7579-20-6
Molecular Formula: C6H6N2O2
Formula Weight: 138.13
Melting Point: ca 280° dec.
MDL No.: MFCD00137840
Formel:
C₆H₆N₂O₂ Molecular Weight: 138,13 g/mol Schmelzpunkt: ∼ 280 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00137840 CAS-Nummer: 7579-20-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...