(tert-Butyldimethylsilyl)acetylen ≥98%

Lieferant: Thermo Scientific
Gefahr

H53494.03 H53494.06 H53494.14
H53494.03EA 70.3 EUR
H53494.03 H53494.06 H53494.14
(tert-Butyldimethylsilyl)acetylen ≥98%
(tert-Butyldimethylsilyl)acetylen

Purity: 98% CAS No.: 86318-61-8 Molecular Formula: C8H16Si Formula Weight: 140.30 Flash Point: 6°(43°F) Refractive Index: 1.451 Boiling Point: 130° Beilstein Reference: 1921466 UN No.: 1993 MDL No.: MFCD00191877

Formel: C₈H₁₆Si
Molecular Weight: 140,3 g/mol
Siedepunkt: 130 °C
Dichte: 0,751
Flammpunkt: 6 °C (43 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00191877
CAS-Nummer: 86318-61-8
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR