Thallium(I)bromid, ultra trocken ≥99,998% (Metall-Basis)
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Thalliumbromid
Unter Argon in Ampullen verpackt
35720.09
35720.18
35720.09EA
153
EUR
35720.09
35720.18
Thallium(I)bromid, ultra trocken ≥99,998% (Metall-Basis)
Thallium(I)bromid
Purity: 99.998%
CAS No.: 7789-40-4
Molecular Formula: TlBr
Formula Weight: 284.28
Physical Form: -10 Mesh Beads, Ampouled under argon
Density: 7.5
Melting Point: ca 460°
Merck Reference: 14,9258
UN No.: 1707
MDL No.: MFCD00011273
Formel:
BrTl Molecular Weight: 284,29 g/mol Siedepunkt: 815 °C (1013 hPa) Schmelzpunkt: ∼460 °C Dichte: 7,5 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011273 CAS-Nummer: 7789-40-4 EINECS: 232-163-0 UN: 1707 ADR: 6.1,II Merck Index: 14,9258 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...