N,N-Dimethylglycinmethylester ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
L11897.06
L11897.14
L11897.06EA
37.5
EUR
L11897.06
L11897.14
N,N-Dimethylglycinmethylester ≥98%
N,N-Dimethylglycinmethylester
Purity: 98%
CAS No.: 7148-06-3
Molecular Formula: C5H11NO2
Formula Weight: 117.15
Flash Point: 38°(100°F)
Refractive Index: 1.4120
Boiling Point: 134-135°
Beilstein Reference: 1744788
UN No.: 3272
MDL No.: MFCD00059303
Formel:
C₅H₁₁NO₂ Siedepunkt: 134…135 °C Dichte: 0,950 Flammpunkt: 38 °C (100 °F) Lagertemperatur: Kühlschrank |
MDL:
MFCD00059303 CAS-Nummer: 7148-06-3 UN: 3272 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...