Tributylphosphit ≥94%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Tri-n-butylphosphite
L14307.AE
L14307.AP
L14307.AEEA
47
EUR
L14307.AE
L14307.AP
Tributylphosphit ≥94%
Tributylphosphit
Purity: 94%
CAS No.: 102-85-2
Molecular Formula: C12H27O3P
Formula Weight: 250.32
Flash Point: 82°(179°F)
Refractive Index: 1.4320
Melting Point: -80°
Boiling Point: 125-128°/10mm
Beilstein Reference: 1703866
MDL No.: MFCD00009437
Formel:
C₁₂H₂₇O₃P Molecular Weight: 250.32 g/mol Siedepunkt: 125…128 °C (10 mmHg) Schmelzpunkt: –80 °C Dichte: 0.915 Flammpunkt: 82 °C (179 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009437 CAS-Nummer: 102-85-2 EINECS: 203-061-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...