3-Fluorisonicotinsäure ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3-Fluoropyridine-4-carboxylic acid

L20042.03 L20042.06
L20042.03EA 27.7 EUR
L20042.03 L20042.06
3-Fluorisonicotinsäure ≥97%
3-Fluorisonicotinsäure

Purity: 97% CAS No.: 393-53-3 Molecular Formula: C6H4FNO2 Formula Weight: 141.10 Melting Point: 251-254° dec. Beilstein Reference: 383678

Formel: C₆H₄FNO₂
Molecular Weight: 141,1 g/mol
Schmelzpunkt: 251…254 °C (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD03789251
CAS-Nummer: 393-53-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR