Methyl-5-brom-2-hydroxynicotinat

Lieferant: BIONET RESEARCH

Synonyme: Methyl 5-bromo-2-oxo-1,2-dihydropyridine-3-carboxylate, methyl 5-bromo-2-hydroxypyridine-3-carboxylate, Methyl 5-bromo-2-oxo-1,2-dihydro-3-pyridinecarboxylate

10W-0208.500 10W-0208.1 10W-0208.5 10W-0208.10
BIOT10W-0208.500EA 0 EUR
BIOT10W-0208.500 BIOT10W-0208.1 BIOT10W-0208.5 BIOT10W-0208.10
Methyl-5-brom-2-hydroxynicotinat
Methyl-5-brom-2-hydroxynicotinat
Formel: C₇H₆BrNO₃
Molecular Weight: 232,03 g/mol
MDL: MFCD03844843
CAS-Nummer: 120034-05-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR