Methyl-5-brom-2-chlornicotinat

Lieferant: BIONET RESEARCH

Synonyme: Methyl 5-bromo-2-chloronicotinate

10W-0710.500 10W-0710.1 10W-0710.5 10W-0710.10
BIOT10W-0710.500EA 0 EUR
BIOT10W-0710.500 BIOT10W-0710.1 BIOT10W-0710.5 BIOT10W-0710.10
Methyl-5-brom-2-chlornicotinat
Methyl-5-brom-2-chlornicotinat
Formel: C₇H₅BrClNO₂
Molecular Weight: 250,48 g/mol
MDL: MFCD03844848
CAS-Nummer: 78686-79-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR