Methyl-6-aminonicotinat

Lieferant: BIONET RESEARCH

Synonyme: Methyl 6-aminonicotinate, 6-Aminonicotinic acid methyl ester

12N-057.500 12N-057.1 12N-057.5 12N-057.10
BIOT12N-057.500EA 0 EUR
BIOT12N-057.500 BIOT12N-057.1 BIOT12N-057.5 BIOT12N-057.10
Methyl-6-aminonicotinat
Methyl-6-aminonicotinat
Formel: C₇H₈N₂O₂
Molecular Weight: 152.15 g/mol
MDL: MFCD00797844
CAS-Nummer: 36052-24-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR