3-Methyl-4-phenyl-1H-pyrazol

Lieferant: BIONET RESEARCH

12T-0222.500 12T-0222.1 12T-0222.5 12T-0222.10
BIOT12T-0222.500EA 0 EUR
BIOT12T-0222.500 BIOT12T-0222.1 BIOT12T-0222.5 BIOT12T-0222.10
3-Methyl-4-phenyl-1H-pyrazol
3-Methyl-4-phenyl-1H-pyrazole
Formel: C₁₀H₁₀N₂
Molecular Weight: 158.2 g/mol
MDL: MFCD00051752
CAS-Nummer: 13788-84-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR