Methyl-2-amino-5-bromnicotinat

Lieferant: BIONET RESEARCH

12W-0701.500 12W-0701.1 12W-0701.5
BIOT12W-0701.500EA 0 EUR
BIOT12W-0701.500 BIOT12W-0701.1 BIOT12W-0701.5
Methyl-2-amino-5-bromnicotinat
Methyl-2-amino-5-bromnicotinat
Formel: C₇H₇BrN₂O₂
MDL: MFCD04117787
CAS-Nummer: 50735-34-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR