(1-Ethoxyethyliden)malononitril

Lieferant: BIONET RESEARCH

Synonyme: 2-(1-Ethoxyethylidene)malononitrile, 2-(1-Ethoxy-ethylidene)-malononitrile

2K-004.500 2K-004.1 2K-004.5 2K-004.10
BIOT2K-004.500EA 0 EUR
BIOT2K-004.500 BIOT2K-004.1 BIOT2K-004.5 BIOT2K-004.10
(1-Ethoxyethyliden)malononitril
(1-Ethoxyethyliden)malononitril
Formel: C₇H₈N₂O
Molecular Weight: 136,15 g/mol
MDL: MFCD00001852
CAS-Nummer: 5417-82-3
EINECS: 226-521-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR