Methyl-5,6-dichlornicotinat

Lieferant: BIONET RESEARCH

Synonyme: Methyl 5,6-dichloronicotinate

3W-0272.500 3W-0272.1 3W-0272.5 3W-0272.10
BIOT3W-0272.500EA 0 EUR
BIOT3W-0272.500 BIOT3W-0272.1 BIOT3W-0272.5 BIOT3W-0272.10
Methyl-5,6-dichlornicotinat
Methyl-5,6-dichlornicotinat
Formel: C₇H₅Cl₂NO₂
MDL: MFCD00125367
CAS-Nummer: 56055-54-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR