3-Aminoisonicotinsäure

Lieferant: BIONET RESEARCH

Synonyme: 3-Aminoisonicotinic Acid, 3-Aminopyridine-4-carboxylic acid

4K-902.500 4K-902.5 4K-902.1
BIOT4K-902.500EA 0 EUR
BIOT4K-902.500 BIOT4K-902.5 BIOT4K-902.1
3-Aminoisonicotinsäure
3-Aminoisonicotinsäure
Formel: C₆H₆N₂O₂
Molecular Weight: 138.13 g/mol
MDL: MFCD00137840
CAS-Nummer: 7579-20-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR