Methyl-6-hydroxynicotinat
Lieferant: BIONET RESEARCH
Synonyme:
Methyl 6-oxo-1,6-dihydro-3-pyridinecarboxylate, Methyl 6-hydroxynicotinate
5E-367S.500
5E-367S.1
5E-367S.5
5E-367S.10
BIOT5E-367S.500EA
0
EUR
BIOT5E-367S.500
BIOT5E-367S.1
BIOT5E-367S.5
BIOT5E-367S.10
Methyl-6-hydroxynicotinat
Methyl-6-hydroxynicotinat
Formel:
C₇H₇NO₃ Molecular Weight: 153,14 g/mol |
MDL:
MFCD04113616 CAS-Nummer: 66171-50-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...