Nicotinaldehydoxim 95

Lieferant: BIONET RESEARCH

Synonyme: 3-Pyridinealdoxime, Pyridine-3-carboxaldoxime, Pyridine-3-aldoxime

5P-059.500 5P-059.1 5P-059.5 5P-059.10
BIOT5P-059.500EA 0 EUR
BIOT5P-059.500 BIOT5P-059.1 BIOT5P-059.5 BIOT5P-059.10
Nicotinaldehydoxim 95
Nicotinaldehydoxim
Formel: C₆H₆N₂O
Molecular Weight: 122.13 g/mol
MDL: MFCD00006409
CAS-Nummer: 1193-92-6
EINECS: 214-782-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR