Methyl-4,6-dichlornicotinat

Lieferant: BIONET RESEARCH

Synonyme: Methyl 4,6-dichloronicotinate

5X-0801.1 5X-0801.5 5X-0801.500
BIOT5X-0801.1EA 0 EUR
BIOT5X-0801.1 BIOT5X-0801.5 BIOT5X-0801.500
Methyl-4,6-dichlornicotinat
Methyl-4,6-dichlornicotinat
Formel: C₇H₅Cl₂NO₂
Molecular Weight: 206,03 g/mol
MDL: MFCD04125732
CAS-Nummer: 65973-52-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR