(4-Methoxybenzyl)malononitril

Lieferant: BIONET RESEARCH

6K-004.500 6K-004.1 6K-004.5 6K-004.10
BIOT6K-004.500EA 0 EUR
BIOT6K-004.500 BIOT6K-004.1 BIOT6K-004.5 BIOT6K-004.10
(4-Methoxybenzyl)malononitril
(4-Methoxybenzyl)malononitril
Formel: C₁₁H₁₀N₂O
MDL: MFCD00231623
CAS-Nummer: 5553-92-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR