Methyl-5-brom-6-chlornicotinat
Lieferant: BIONET RESEARCH
Synonyme:
Methyl 5-bromo-6-chloronicotinate
7W-0337.500
7W-0337.1
7W-0337.5
7W-0337.10
BIOT7W-0337.500EA
0
EUR
BIOT7W-0337.500
BIOT7W-0337.1
BIOT7W-0337.5
BIOT7W-0337.10
Methyl-5-brom-6-chlornicotinat
Methyl-5-brom-6-chlornicotinat
Formel:
C₇H₅BrClNO₂ Molecular Weight: 250.48 g/mol |
MDL:
MFCD03844855 CAS-Nummer: 78686-77-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...