Methyl-2,5-dichlornicotinat
Lieferant: BIONET RESEARCH
Synonyme:
Methyl 2,5-dichloronicotinate
7W-0397.500
7W-0397.1
7W-0397.5
7W-0397.10
BIOT7W-0397.500EA
0
EUR
BIOT7W-0397.500
BIOT7W-0397.1
BIOT7W-0397.5
BIOT7W-0397.10
Methyl-2,5-dichlornicotinat
Methyl-2,5-dichlornicotinat
Formel:
C₇H₅Cl₂NO₂ Molecular Weight: 206,03 g/mol |
MDL:
MFCD03844857 CAS-Nummer: 67754-03-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...