Methyl-2-chlor-5-iodnicotinat
Lieferant: BIONET RESEARCH
Synonyme:
Methyl 2-chloro-5-iodonicotinate, Methyl 2-chloro-5-iodopyridine-3-carboxylate
9Y-0700.1
9Y-0700.500
BIOT9Y-0700.1EA
0
EUR
BIOT9Y-0700.1
BIOT9Y-0700.500
Methyl-2-chlor-5-iodnicotinat
Methyl-2-chlor-5-iodnicotinat
Formel:
C₇H₅ClINO₂ Molecular Weight: 297.48 g/mol |
MDL:
MFCD06659607 CAS-Nummer: 78686-83-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...