(R)-(-)-β-Aminophenethylalkohol
Lieferant: Cayman Chemical
Synonyme:
D-Phenylglycinol, (R)-(-)-2-Phenylglycinol
10007132-1
10007132-10
10007132-25
10007132-5
CAYM10007132-1EA
28.6
EUR
CAYM10007132-1
CAYM10007132-10
CAYM10007132-25
CAYM10007132-5
(R)-(-)-β-Aminophenethylalkohol
(R)-(-)-β-Aminophenethylalkohol
Formel:
C₈H₁₁NO Molecular Weight: 137,18 g/mol |
MDL:
MFCD00008062 CAS-Nummer: 56613-80-0 EINECS: 260-287-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...