Aminoguanidin Hydrochlorid
Lieferant: Cayman Chemical
Synonyme:
Aminoguanidine HCl, Aminoguanidine Hydrochloride, Guanylhydrazine hydrochloride
81530-10
81530-100
81530-25
81530-50
CAYM81530-10EA
34
EUR
CAYM81530-10
CAYM81530-100
CAYM81530-25
CAYM81530-50
Aminoguanidin Hydrochlorid
Aminoguanidin Hydrochlorid
Formel:
CH₇ClN₄ |
MDL:
MFCD00039074 CAS-Nummer: 1937-19-5 EINECS: 217-707-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...