4-(4-Nitrobenzyloxy)benzaldehyd

Lieferant: CHEMPUR

BTB09464.250 BTB09464.1
CHMPBTB09464.250EA 0 EUR
CHMPBTB09464.250 MAYBBTB09464.1
4-(4-Nitrobenzyloxy)benzaldehyd
4-(4-Nitrobenzyloxy)benzaldehyd
Formel: C₁₄H₁₁NO₄
Molecular Weight: 257.25 g/mol
MDL: MFCD00017041
CAS-Nummer: 67565-48-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR