3-Methyl-4-phenyl-1H-pyrazol

Lieferant: Thermo Scientific

CD09292.1 CD09292.10 CD09292.50
MAYBCD09292.1EA 0 EUR
MAYBCD09292.1 MAYBCD09292.10 MAYBCD09292.50
3-Methyl-4-phenyl-1H-pyrazol
3-Methyl-4-phenyl-1H-pyrazole
Formel: C₁₀H₁₀N₂
MDL: MFCD00051752
CAS-Nummer: 13788-84-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR