Methyl-5-methylnicotinat

Lieferant: CHEMPUR

Synonyme: Methyl 5-methylnicotinate

KM10012.250 KM10012.1
CHMPKM10012.250EA 0 EUR
CHMPKM10012.250 MAYBKM10012.1
Methyl-5-methylnicotinat
Methyl-5-methylnicotinat
Formel: C₈H₉NO₂
Molecular Weight: 151,16 g/mol
MDL: MFCD01075739
CAS-Nummer: 29681-45-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR