N-Ethylacetamid

Lieferant: CHEMPUR

KM10028.250 KM10028.1
CHMPKM10028.250EA 0 EUR
CHMPKM10028.250 MAYBKM10028.1
N-Ethylacetamid
N-Ethylacetamid
Formel: C₄H₉NO
Molecular Weight: 87,12 g/mol
MDL: MFCD00009029
CAS-Nummer: 625-50-3
EINECS: 210-896-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR