4-Methoxy-3-nitrobenzaldehyd

Lieferant: CHEMPUR

NRB03381.250 NRB03381.1
CHMPNRB03381.250EA 0 EUR
CHMPNRB03381.250 MAYBNRB03381.1
4-Methoxy-3-nitrobenzaldehyd
4-Methoxy-3-nitrobenzaldehyde
Formel: C₈H₇NO₄
Molecular Weight: 181,15 g/mol
MDL: MFCD00126498
CAS-Nummer: 31680-08-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR