2-Chlor-6-methoxyisonicotinsäure

Lieferant: Thermo Scientific

Synonyme: 2-Chloro-6-methoxypyridine-4-carboxylic acid

RF03693DA RF03693EA
MAYBRF03693DAEA 0 EUR
MAYBRF03693DA MAYBRF03693EA
2-Chlor-6-methoxyisonicotinsäure
2-Chlor-6-methoxyisonicotinsäure
Formel: C₇H₆ClNO₃
Molecular Weight: 187.58 g/mol
MDL: MFCD00173928
CAS-Nummer: 15855-06-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR