6-Chlorpicolinsäure
Lieferant: Thermo Scientific
Synonyme:
6-Chloropyridine-2-carboxylic acid, 2-Chloropyridine-6-carboxylic acid
RJC02242.250
RJC02242.1
MAYBRJC02242.250EA
0
EUR
MAYBRJC02242.250
MAYBRJC02242.1
6-Chlorpicolinsäure
6-Chlorpicolinsäure
Formel:
C₆H₄ClNO₂ Molecular Weight: 157,56 g/mol |
MDL:
MFCD00155390 CAS-Nummer: 4684-94-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...