Cysteaminiumchlorid

Lieferant: Thermo Scientific

Synonyme: 2-MEA

SB00733.10 SB00733.50
MAYBSB00733.10EA 0 EUR
MAYBSB00733.10 MAYBSB00733.50
Cysteaminiumchlorid
Cysteaminiumchlorid
Formel: C₂H₇NS·HCl
Molecular Weight: 113.61 g/mol
MDL: MFCD00012904
CAS-Nummer: 156-57-0
EINECS: 205-858-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR