3-Chinuclidinon Hydrochlorid

Lieferant: Thermo Scientific

Synonyme: 1-Azabicyclo[2.2.2]octan-3-one hydrochloride

SB01126.1 SB01126.10 SB01126.50
MAYBSB01126.1EA 0 EUR
MAYBSB01126.1 MAYBSB01126.10 MAYBSB01126.50
3-Chinuclidinon Hydrochlorid
3-Chinuclidinon Hydrochlorid
Formel: C₇H₁₂ClNO
Molecular Weight: 161.63 g/mol
MDL: MFCD00137391
CAS-Nummer: 1193-65-3
EINECS: 214-776-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR